Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou6f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101002.1 | NP_001094472.1 | 8 | 1554 | ||
GenBank mRNA | BC128774.1 | - | 8 | 1554 | 7132745 |