Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meiob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115033.1 | NP_001108505.1 | 7 | 628 | ||
XM_006246031.2 | XP_006246093.1 | 7 | 843 | |||
XM_008767593.1 | XP_008765815.1 | 7 | 715 | |||
GenBank mRNA | BC158604.1 | - | 7 | 628 | 7134815 |