Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115036.1 | NP_001108508.1 | 2 | 200 | ||
XM_006239067.2 | XP_006239129.1 | 3 | 319 | |||
XM_006239068.2 | XP_006239130.1 | 2 | 304 | |||
GenBank mRNA | BC107929.1 | - | 2 | 206 | ||
BC158610.1 | - | 2 | 179 | 7374180 | ||
FQ225384.1 | - | 2 | 199 | |||
FQ232337.1 | - | 2 | 199 |