Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115036.1 | NP_001108508.1 | 3, 4 | 416 | ||
XM_006239067.2 | XP_006239129.1 | 4, 5 | 535 | |||
XM_006239068.2 | XP_006239130.1 | 3, 4 | 520 | |||
GenBank mRNA | BC107929.1 | - | 4, 5 | 550 | ||
BC158610.1 | - | 3, 4 | 395 | 7374180 | ||
FQ225384.1 | - | 3, 4 | 415 | |||
FQ232337.1 | - | 3, 4 | 415 |