Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204053.1 | NP_001190982.1 | 2, 3 | 533 | ||
GenBank mRNA | FQ086998.1 | - | 2, 3 | 533 |