Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127600.1 | NP_001121072.1 | 6 | 529 | ||
GenBank mRNA | BC161899.1 | - | 6 | 529 | 8367117 |