Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ddr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031764.3 | NP_113952.2 | 10 | 1399 | ||
XM_006250226.2 | XP_006250288.1 | 11 | 4696 | |||
XM_006250227.2 | XP_006250289.1 | 13 | 2031 | |||
XM_006250228.2 | XP_006250290.1 | 12 | 1812 | |||
XM_006250229.2 | XP_006250291.1 | 12 | 1760 | |||
XM_008769761.1 | XP_008767983.1 | 12 | 1774 | |||
XM_008769762.1 | XP_008767984.1 | 12 | 1706 | |||
XM_008769763.1 | XP_008767985.1 | 12 | 2012 | |||
GenBank mRNA | BC161961.1 | - | 10 | 1399 | 9027893 | |
BQ207033.1 | - | 3 | 461 |