Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127570.1 | NP_001121042.1 | 8 | 1405 | ||
GenBank mRNA | BC161996.1 | - | 8 | 1405 | 9034684 |