Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031059.1 | NP_112321.1 | 2 | 1020 | ||
GenBank mRNA | D83036.1 | - | 2 | 1020 |