Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hip1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134763.1 | NP_001128235.1 | 21 | 2138 | ||
NM_031234.1 | NP_112513.1 | 21 | 2135 | |||
XM_008769266.1 | XP_008767488.1 | 21 | 2248 | |||
XM_008769267.1 | XP_008767489.1 | 21 | 2245 | |||
XM_008769268.1 | XP_008767490.1 | 21 | 4298 | |||
GenBank mRNA | AB005052.1 | - | 21 | 2132 | ||
BC169096.1 | - | 21 | 2122 | 7936362 |