Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031329.2 | NP_112619.2 | 7, 8 | 1574 | ||
XM_006231853.2 | XP_006231915.1 | 7, 8 | 1570 | |||
XM_006231854.2 | XP_006231916.1 | 7, 8 | 1614 | |||
XM_006231855.2 | XP_006231917.1 | 6, 7 | 1494 | |||
GenBank mRNA | BC062037.1 | - | 7, 8 | 1574 | 5623744 |