Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031828.1 | NP_114016.1 | 24 | 3044 | ||
GenBank mRNA | AF083341.1 | - | 22 | 2297 | ||
AF135265.1 | - | 22 | 2715 | |||
AY330290.1 | - | 21 | 2174 | |||
AY330291.1 | - | 21 | 2174 | |||
AY330292.1 | - | 21 | 2183 | |||
AY330293.2 | - | 21 | 2174 | |||
AY330294.1 | - | 21 | 2174 | |||
AY344964.1 | - | 19 | 2010 | |||
AY344965.2 | - | 20 | 2586 | |||
U40603.1 | - | 24 | 3044 | |||
U55995.1 | - | 21 | 2327 | |||
U93052.1 | - | 21 | 2275 |