Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110333.2 | NP_001103803.1 | 3 | 1217 | ||
NM_001110334.2 | NP_001103804.1 | 3 | 1217 | |||
NM_001287107.1 | NP_001274036.1 | 3 | 1217 | |||
NM_001287108.1 | NP_001274037.1 | 3 | 1217 | |||
NM_001287110.1 | NP_001274039.1 | 3 | 1217 | |||
NM_001287111.1 | NP_001274040.1 | 3 | 1217 | |||
NM_001287112.1 | NP_001274041.1 | 3 | 1217 | |||
NM_001287113.1 | NP_001274042.1 | 2 | 1168 | |||
NM_001287114.1 | NP_001274043.1 | 2 | 1168 | |||
NM_031836.3 | NP_114024.2 | 3 | 1217 | |||
GenBank mRNA | AF080594.1 | - | 2 | 125 | ||
AF215725.1 | - | 3 | 203 | |||
AF215726.1 | - | 3 | 203 | |||
AF222779.1 | - | 3 | 203 | |||
AY033503.1 | - | 2 | 165 | |||
AY033504.1 | - | 2 | 165 | |||
AY033506.1 | - | 3 | 214 | |||
AY033507.1 | - | 3 | 214 | |||
AY033508.1 | - | 3 | 214 | |||
AY702972.1 | - | 3 | 203 | |||
BC168708.1 | - | 3 | 1093 | 7458665 | ||
GQ423618.1 | - | 2 | 125 | |||
M32167.1 | - | 3 | 207 |