Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031979.3 | NP_114185.3 | 6, 7 | 937 | ||
GenBank mRNA | BC105778.1 | - | 6, 7 | 932 | 7932542 | |
U22893.1 | - | 6, 7 | 940 |