Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053419.1 | NP_445871.1 | 10 | 1245 | ||
GenBank mRNA | AF169549.1 | - | 3 | 216 | ||
AF242544.1 | - | 8 | 903 | |||
EF121977.1 | - | 8 | 708 | |||
EF121978.1 | - | 8 | 708 | |||
EF121979.1 | - | 9 | 942 | |||
EF121980.1 | - | 9 | 942 | |||
EF121981.1 | - | 10 | 1245 | |||
EF121982.1 | - | 8 | 708 |