Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109907.1 | NP_001103377.1 | 9, 10 | 1482 | ||
NM_053436.2 | NP_445888.1 | 9, 10 | 1500 | |||
GenBank mRNA | AF227200.1 | - | 9, 10 | 1487 | ||
AF290989.1 | - | 9, 10 | 1469 | |||
BC081755.1 | - | 9, 10 | 1440 | |||
BC101858.1 | - | 9, 10 | 1478 | 7937343 |