Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053586.1 | NP_446038.1 | 2, 3 | 197 | ||
GenBank mRNA | BC083179.1 | - | 2, 3 | 210 | 7300877 | |
D10952.1 | - | 2, 3 | 197 | |||
FQ210284.1 | - | 2, 3 | 194 | |||
FQ214227.1 | - | 2, 3 | 200 | |||
FQ215884.1 | - | 2, 3 | 200 | |||
FQ217498.1 | - | 2, 3 | 200 | |||
FQ217781.1 | - | 2, 3 | 218 | |||
FQ217835.1 | - | 2, 3 | 200 | |||
FQ218282.1 | - | 2, 3 | 198 | |||
FQ224825.1 | - | 2, 3 | 211 | |||
X14208.1 | - | 2, 3 | 145 |