Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020529.2 | NP_065390.1 | 3 | 532 | ||
GenBank mRNA | AK311515.1 | - | 2 | 294 | ||
AK313421.1 | - | 3 | 528 | |||
AY033600.1 | - | 3 | 436 | |||
BC002601.2 | - | 3 | 524 | 3161686 | ||
BC004983.1 | - | 3 | 515 | 2957970 | ||
BT007091.1 | - | 3 | 422 | |||
M69043.1 | - | 3 | 516 |