Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
siRNA ID
VHS40700
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
4
467
NM_001318068.1
NP_001304997.1
5
621
NM_001318069.1
NP_001304998.1
5
1032
NM_002753.4
NP_002744.1
5
568
NM_138980.3
NP_620446.1
5
1059
NM_138982.3
NP_620448.1
5
1032
XM_005263129.2
XP_005263186.1
5
1129
XM_005263130.2
XP_005263187.1
5
525
XM_005263131.2
XP_005263188.1
5
926
XM_005263135.3
XP_005263192.1
5
926
XM_006714268.2
XP_006714331.1
5
799
XM_006714269.2
XP_006714332.1
5
926
XM_011532117.2
XP_011530419.1
5
632
XM_011532118.2
XP_011530420.1
5
725
XM_011532120.2
XP_011530422.1
5
695
XM_011532121.2
XP_011530423.1
5
561
XM_017008419.1
XP_016863908.1
6
822
XM_017008420.1
XP_016863909.1
5
1256
XM_017008421.1
XP_016863910.1
5
852
XM_017008422.1
XP_016863911.1
3
352
XM_017008423.1
XP_016863912.1
5
544
XM_017008424.1
XP_016863913.1
5
879
XM_017008425.1
XP_016863914.1
5
552
XM_017008426.1
XP_016863915.1
5
880
XM_017008427.1
XP_016863916.1
5
953
XM_017008428.1
XP_016863917.1
5
695
XM_017008429.1
XP_016863918.1
5
1256
XM_017008430.1
XP_016863919.1
5
634
XM_017008431.1
XP_016863920.1
5
852
XM_017008432.1
XP_016863921.1
5
525
XM_017008433.1
XP_016863922.1
5
953
XM_017008434.1
XP_016863923.1
5
909
XM_017008435.1
XP_016863924.1
5
799
XM_017008436.1
XP_016863925.1
5
1283
XM_017008437.1
XP_016863926.1
5
561
XM_017008438.1
XP_016863927.1
5
552
XM_017008439.1
XP_016863928.1
5
880
XM_017008440.1
XP_016863929.1
5
953
XM_017008441.1
XP_016863930.1
5
953
XM_017008442.1
XP_016863931.1
7
1274
XM_017008443.1
XP_016863932.1
7
925
XM_017008444.1
XP_016863933.1
7
1247
XM_017008445.1
XP_016863934.1
4
756
XM_017008446.1
XP_016863935.1
2
1448
XM_017008447.1
XP_016863936.1
5
953
XM_017008448.1
XP_016863937.1
5
926
XM_017008449.1
XP_016863938.1
5
953
XM_017008450.1
XP_016863939.1
5
926
XM_017008451.1
XP_016863940.1
4
693
XM_017008452.1
XP_016863941.1
5
953
XM_017008453.1
XP_016863942.1
3
352
XM_017008454.1
XP_016863943.1
5
632
GenBank mRNA
AK057723.1
-
1
413
AK124791.1
-
5
1284
AK225974.1
-
5
613
AK294125.1
-
1
83
AK307129.1
-
6
745
BC022492.1
-
5
865
BC035057.1
-
5
877
5261221
BC051731.1
-
6
737
4838495
BC065516.1
-
1
88
5721954
U07620.1
-
5
569
U34819.1
-
4
413
U34820.1
-
4
437
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0