Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPS6KA3 SIRNA Assays ›
Gene Symbol:
RPS6KA3
Gene Name:
ribosomal protein S6 kinase A3
Gene Aliases:
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
Chromosome Location:
Chr.X: 20149911 - 20267514 on Build GRCh38
UniGene ID:
Hs.445387
Species:
Human
siRNA ID
VHS40856
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RPS6KA3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004586.2
NP_004577.1
7
542
XM_005274573.2
XP_005274630.1
7
546
XM_005274577.2
XP_005274634.1
6
482
XM_006724507.3
XP_006724570.1
9
648
XM_011545555.1
XP_011543857.1
7
546
XM_011545556.1
XP_011543858.1
7
546
XM_011545557.2
XP_011543859.1
9
664
XM_011545558.2
XP_011543860.1
9
728
XM_011545559.1
XP_011543861.1
9
725
XM_011545560.1
XP_011543862.1
8
580
XM_011545561.1
XP_011543863.1
7
506
XM_011545562.1
XP_011543864.1
6
482
XM_011545563.2
XP_011543865.1
9
662
XM_017029713.1
XP_016885202.1
9
650
XM_017029714.1
XP_016885203.1
9
726
XM_017029715.1
XP_016885204.1
9
725
XM_017029716.1
XP_016885205.1
9
800
XM_017029717.1
XP_016885206.1
7
506
XM_017029718.1
XP_016885207.1
6
482
XM_017029719.1
XP_016885208.1
7
506
GenBank mRNA
AB102662.1
-
7
501
AB208933.1
-
7
744
AK294373.1
-
9
637
AK313932.1
-
7
546
AK316482.1
-
8
941
BC096301.1
-
7
542
40007452
BC096302.3
-
7
542
40007453
BC096303.1
-
7
542
40007456
L07599.1
-
7
539
U08316.1
-
7
542
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
cell cycle
signal transduction
central nervous system development
positive regulation of cell growth
response to lipopolysaccharide
intracellular signal transduction
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of translation in response to stress
regulation of DNA-templated transcription in response to stress
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
protein kinase binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0