Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292039.1 | NP_001278968.1 | 3, 4 | 442 | ||
NM_002747.3 | NP_002738.2 | 4, 5 | 1858 | |||
XM_005258299.3 | XP_005258356.1 | 4, 5 | 6220 | |||
XM_011526074.2 | XP_011524376.1 | 4, 5 | 6402 | |||
XM_011526075.2 | XP_011524377.1 | 4, 5 | 1860 | |||
XM_011526076.2 | XP_011524378.1 | 4, 5 | 2223 | |||
XM_011526077.2 | XP_011524379.1 | 3, 4 | 2869 | |||
XM_017025839.1 | XP_016881328.1 | 4, 5 | 1791 | |||
GenBank mRNA | AK293818.1 | - | 3, 4 | 423 | ||
AK295058.1 | - | 4, 5 | 1779 | |||
AK303601.1 | - | 3, 4 | 520 | |||
BC050299.1 | - | 4, 5 | 1858 | 5303135 | ||
BC117216.1 | - | 3, 4 | 912 | 40125767 | ||
X59727.1 | - | 3, 4 | 1395 |