Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207013.1 | NP_001193942.1 | 4, 5 | 545 | ||
NM_001308287.1 | NP_001295216.1 | 3, 4 | 440 | |||
NM_002828.3 | NP_002819.2 | 4, 5 | 545 | |||
NM_080422.2 | NP_536347.1 | 4, 5 | 545 | |||
NM_080423.2 | NP_536348.1 | 4, 5 | 545 | |||
XM_005258124.3 | XP_005258181.1 | 4, 5 | 543 | |||
XM_005258125.3 | XP_005258182.1 | 4, 5 | 543 | |||
XM_011525705.2 | XP_011524007.1 | 5, 6 | 763 | |||
XM_017025884.1 | XP_016881373.1 | 4, 5 | 561 | |||
XM_017025885.1 | XP_016881374.1 | 5, 6 | 747 | |||
XM_017025886.1 | XP_016881375.1 | 3, 4 | 435 | |||
XM_017025887.1 | XP_016881376.1 | 3, 4 | 386 | |||
XM_017025888.1 | XP_016881377.1 | 3, 4 | 386 | |||
GenBank mRNA | AB209569.1 | - | 3, 4 | 472 | ||
AK225706.1 | - | 4, 5 | 424 | |||
AK290649.1 | - | 4, 5 | 427 | |||
AK292570.1 | - | 4, 5 | 445 | |||
BC008244.1 | - | 4, 5 | 388 | 3872164 | ||
BC016727.1 | - | 4, 5 | 457 | 4081330 | ||
BG699440.1 | - | 4, 5 | 550 | |||
M25393.1 | - | 4, 5 | 411 |