Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278618.1 | NP_001265547.1 | 6 | 839 | ||
NM_002827.3 | NP_002818.1 | 7 | 930 | |||
GenBank mRNA | AK290638.1 | - | 7 | 897 | ||
AK299830.1 | - | 6 | 777 | |||
AK316563.1 | - | 7 | 872 | |||
BC015660.2 | - | 7 | 859 | 4844022 | ||
BC018164.1 | - | 7 | 840 | 4338750 | ||
BT006752.1 | - | 7 | 723 | |||
M31724.1 | - | 7 | 813 | |||
M33689.1 | - | 7 | 795 |