Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 21, 22 | 3058 | ||
NM_001321852.1 | NP_001308781.1 | 20, 21 | 3061 | |||
NM_001321853.1 | NP_001308782.1 | 22, 23 | 3320 | |||
NM_001321854.1 | NP_001308783.1 | 21, 22 | 3236 | |||
NM_001321855.1 | NP_001308784.1 | 21, 22 | 3239 | |||
NM_001321856.1 | NP_001308785.1 | 20, 21 | 2974 | |||
NM_001321857.1 | NP_001308786.1 | 20, 21 | 3152 | |||
NM_002227.3 | NP_002218.2 | 20, 21 | 3155 | |||
XM_017001212.1 | XP_016856701.1 | 20, 21 | 2999 | |||
GenBank mRNA | AB209057.1 | - | 19, 20 | 2911 | ||
AB219242.1 | - | 18, 19 | 2802 | |||
AK314257.1 | - | 21, 22 | 3059 | |||
BC132729.1 | - | 19, 20 | 2873 | 40146751 | ||
BX647675.1 | - | 6, 7 | 759 | |||
HQ628631.1 | - | 18, 19 | 2835 | |||
M64174.1 | - | 19, 20 | 2880 |