Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 5 | 1362 | ||
XM_005264237.3 | XP_005264294.1 | 6 | 1635 | |||
XM_005264238.3 | XP_005264295.1 | 5 | 611 | |||
XM_017003741.1 | XP_016859230.1 | 7 | 1640 | |||
XM_017003742.1 | XP_016859231.1 | 6 | 679 | |||
XM_017003743.1 | XP_016859232.1 | 3 | 361 | |||
XM_017003744.1 | XP_016859233.1 | 2 | 109 | |||
GenBank mRNA | AB015982.2 | - | 5 | 1362 | ||
AK027585.1 | - | 2 | 171 | |||
BC012820.2 | - | 5 | 1321 | |||
BC030706.2 | - | 5 | 1307 | 5266769 |