Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec6a-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002706.1 | 3, 4 | 210 | |||
GenBank mRNA | AY370893.1 | - | 3, 4 | 210 |