Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1564534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028321.1 | 3 | 2353 | |||
GenBank mRNA | BC111405.1 | - | 3 | 2353 | 7446493 |