Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sprr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003548.1 | 2 | 462 | |||
GenBank mRNA | AY158991.1 | - | 2 | 461 | ||
BC130233.1 | - | 2 | 445 |