Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210015D19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015577.1 | 5 | 801 | |||
GenBank mRNA | AK005556.1 | - | 5 | 801 | ||
AK008730.1 | - | 3 | 532 | |||
AK009198.1 | - | 3 | 559 | |||
BC052911.1 | - | 3 | 521 | |||
BC094935.1 | - | 3 | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX238328.1 | - | 3 | 521 | Chr. 11 - 5762166 - 5788324 [+] on Build GRCm38 |