Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029459.1 | 2, 3 | 282 | |||
GenBank mRNA | BC055849.1 | - | 2, 3 | 202 | ||
CJ066501.1 | - | 2, 3 | 220 |