Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb2a-ps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003964.2 | 1 | 108 | |||
GenBank mRNA | BG066514.2 | - | 1 | 108 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | C79445.1 | - | 105 | Chr. - - [-] on Build |