Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs5-8s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003280.2 | 32 | ||||
GenBank mRNA | J01871.1 | - | 33 |
Gene Symbol: Lyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ311501.1 | - | 32 |
Gene Symbol: Rn45s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046233.2 | 6909 |