- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027810.1 | 1 | 511 | |||
GenBank mRNA | AB041800.1 | - | 1 | 510 | ||
AK006095.1 | - | 1 | 481 | |||
BC049589.1 | - | 1 | 490 | |||
BC061108.1 | - | 1 | 501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA463268.1 | - | 1 | 493 | Chr. 5 - 115631111 - 115631795 [-] on Build GRCm38 |