Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027810.1 | 1 | 450 | |||
GenBank mRNA | AB041800.1 | - | 1 | 449 | ||
AK003508.1 | - | 1 | 450 | |||
AK006095.1 | - | 1 | 420 | |||
BC049589.1 | - | 1 | 429 | |||
BC061108.1 | - | 1 | 440 |