- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027810.1 | 1 | 750 | |||
GenBank mRNA | AK003508.1 | - | 1 | 750 | ||
AK006095.1 | - | 1 | 720 | |||
BC049589.1 | - | 1 | 729 | |||
BC061108.1 | - | 1 | 740 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703769.1 | - | 1 | 748 | Chr. 5 - 115631045 - 115631815 [-] on Build GRCm38 |