- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027810.1 | 1 | 660 | |||
GenBank mRNA | AB041800.1 | - | 1 | 659 | ||
AK003508.1 | - | 1 | 660 | |||
AK006095.1 | - | 1 | 630 | |||
BC049589.1 | - | 1 | 639 | |||
BC061108.1 | - | 1 | 650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703769.1 | - | 1 | 658 | Chr. 5 - 115631045 - 115631815 [-] on Build GRCm38 |