Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027810.1 | 1 | 486 | |||
GenBank mRNA | AB041800.1 | - | 1 | 485 | ||
AK003508.1 | - | 1 | 486 | |||
AK006095.1 | - | 1 | 456 | |||
BC049589.1 | - | 1 | 465 | |||
BC061108.1 | - | 1 | 476 |