- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027810.1 | 1 | 629 | |||
GenBank mRNA | AB041800.1 | - | 1 | 628 | ||
AK003508.1 | - | 1 | 629 | |||
AK006095.1 | - | 1 | 599 | |||
BC049589.1 | - | 1 | 608 | |||
BC061108.1 | - | 1 | 619 |