Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn1-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029430.1 | 1 | 803 | |||
GenBank mRNA | CF896624.1 | - | 1 | 546 | ||
CF897466.1 | - | 1 | 516 |