Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027827.1 | 2 | 1100 | |||
GenBank mRNA | AB041807.1 | - | 2 | 1052 | ||
BC021519.1 | - | 2 | 1053 |