Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: E130201H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024324.1 | 1 | 678 | |||
GenBank mRNA | AK021400.1 | - | 1 | 680 | ||
BC094539.1 | - | 1 | 653 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB542603.1 | - | 1 | 132 | Chr. 7 - 120598176 - 120598472 [+] on Build GRCm38 |