Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2010204K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027924.1 | 2 | 381 | |||
NR_027925.1 | 2 | 391 | ||||
GenBank mRNA | AF038507.1 | - | 2 | 345 | ||
AK002502.1 | - | 730 | ||||
AK008437.1 | - | 2 | 382 | |||
AK077356.1 | - | 2 | 425 | |||
BC042200.1 | - | 2 | 344 | |||
BC061056.1 | - | 2 | 392 | |||
BC072610.1 | - | 2 | 305 |