Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015385.2 | 2 | 487 | |||
NR_015386.2 | 2 | 396 | ||||
NR_015387.2 | 3 | 749 | ||||
NR_038082.1 | 2 | 396 | ||||
NR_038083.1 | 2 | 396 | ||||
NR_038084.1 | 2 | 396 | ||||
NR_038085.1 | 2 | 396 | ||||
NR_038086.1 | 2 | 396 | ||||
GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
2 | 2258 | |
AK053722.1 | - | 2 | 398 | |||
AY589790.1 | - | 2 | 275 | |||
AY589791.1 | - | 3 | 718 | |||
AY589792.1 | - | 1411 | ||||
AY590888.1 | - | 2 | 225 | |||
AY590889.1 | - | 2 | 315 | |||
BC065087.1 | - | 2 | 385 | |||
CA527570.1 | - | 2 | 340 | |||
CF737515.1 | - | 2 | 356 | |||
CF737925.1 | - | 2 | 372 |