Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox2ot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015580.2 | 5 | 2237 | |||
GenBank mRNA | AK031826.1 | - | 2 | 1797 | ||
AK045614.1 | - | 3 | 1973 | |||
BC057611.1 | - | 5 | 2238 |