Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_029384.1 | 5 | 704 | |||
GenBank mRNA | AK042665.1 | - | LIT3409 Otx2OS |
5 | 704 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB246370.2 | - | 5 | 666 | Chr. 14 - 48669326 - 48793345 [+] on Build GRCm38 |