Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033304.2 | 2 | 221 | |||
GenBank mRNA | AK053620.1 | - | 2 | 187 | ||
BC070462.1 | - | 1 | 77 | 30650880 |