Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vax2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY589787.1 | - | LIT3407 Vax2OS |
2 | 465 |