Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A630089N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015491.1 | 4, 5 | 409 | |||
GenBank mRNA | AK041610.1 | - | 4, 5 | 421 | ||
AK042413.1 | - | 4, 5 | 410 |