Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A630089N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015491.1 | 5 | 1777 | |||
GenBank mRNA | AK042413.1 | - | 5 | 1778 |