Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A630089N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015491.1 | 5 | 1807 | |||
GenBank mRNA | AK042413.1 | - | 5 | 1808 |
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141097.1 | - | 7 | 1544 |